Kimble™ KIMAX™ Flasks with Reinforced Beaded Top
Kimble™ KIMAX™ Flasks with Reinforced Beaded Top
Actual product may vary
Fisherbrand
Kimble™ KIMAX™ Flasks with Reinforced Beaded Top
Tooled heavy-duty top finish
Have Questions?
Catalog NumberCapacity (Metric)
FIS10-039M2000 mL
Catalog number FIS10-039M
Price (MYR)
1,010.00
PK
Add to cart
Capacity (Metric):
2000 mL
Price (MYR)
1,010.00
PK
Add to cart
Tooled heavy-duty top finish
  • Durable white enamel capacity scale
  • Uniformly thick walls
  • Long tapered outside contour
  • 250, 300, and 500mL sizes can use covers in place of stoppers
  • Stoppers not included
Specifications
Closure MaterialRubber
DescriptionWhite Ceramic Enamel capacity scale; Top style: Reinforced and tooled with rounded finish beaded
Diameter (English) Outer6.33 in.
Capacity (Metric)2000 mL
MaterialBorosilicate Glass
Graduation Range600 to 2000 mL
Graduations100 mL
Height (English)10.82 in.
Height (Metric)27.5 cm
Mouth StyleNarrow
Diameter (Metric) Outer160 mm
Closure Size10
Unit SizePK