Check Order Status
Rxi™ Guard/Retention Gap Columns
Actual product may vary
Fisherbrand
Rxi™ Guard/Retention Gap Columns
Extend column lifetime
Have Questions?
Catalog Number
Length (Metric)
Diameter (Metric) Inner
Quantity
FIS10-029
5 m
0.25 mm
1/Pk.
Catalog number
FIS10-029
Price (MYR)
605.00
EA
Add to cart
Length (Metric):
5 m
Diameter (Metric) Inner:
0.25 mm
Quantity:
1/Pk.
Price (MYR)
605.00
EA
Add to cart
Extends column lifetime
Excellent inertness achieves lower detection limits for active compounds
Sharper chromatographic peaks by utilizing retention gap technology
Maximum temperature: 360°C
Specifications
For Use With (Equipment)
Rxi™ Columns
Type
Guard/Retention Gap Columns
Diameter (Metric) Inner
0.25 mm
Length (Metric)
5 m
Material
Fused Silica
Quantity
1/Pk.