Check Order Status
PYREX™ Short Neck Flat Bottom Flasks, Standard Taper Joint
Actual product may vary
Fisherbrand
PYREX™ Short Neck Flat Bottom Flasks, Standard Taper Joint
Short Necks
Have Questions?
Catalog Number
Capacity (Metric)
FIS09-559A
125 mL
Catalog number
FIS09-559A
Price (MYR)
352.00
EA
Add to cart
Capacity (Metric):
125 mL
Price (MYR)
352.00
EA
Add to cart
Short Necks
With full length outer joints, but with shorter necks.
Specifications
Diameter (English) Outer
2.7 in.
Capacity (Metric)
125 mL
Material
Borosilicate Glass
Height (English)
4.7 in.
Height (Metric)
12 cm
Neck Style
Short
No. per Case
12/Cs.
No. per Pack
12/Pk.
Standard Taper Joint
24/40
Disposable
Reusable
Bottom Shape
Flat
Diameter (Metric) Outer
69 mm