PYREX™ Kjeldahl Nitrogen Distilling Apparatus, Standard Taper Joints
PYREX™ Kjeldahl Nitrogen Distilling Apparatus, Standard Taper Joints
Actual product may vary
Fisherbrand
PYREX™ Kjeldahl Nitrogen Distilling Apparatus, Standard Taper Joints
Determines nitrogen in organic compounds
Have Questions?
Catalog NumberType
FIS09-128Kjeldahl Nitrogen Distillation Apparatus
Catalog number FIS09-128
Price (MYR)
5,006.00
CS
Add to cart
Type:
Kjeldahl Nitrogen Distillation Apparatus
Price (MYR)
5,006.00
CS
Add to cart
This PYREX™ 500 mL distilling apparatus is used for determining nitrogen in organic compounds. This assembly includes a Kjeldahl flask specifically developed for this process and a Graham condenser with 10mm O.D. tubulations. Both the digestion and distillation stages can be performed in this flask. The assembly shown is the distillation stage, where the amount of ammonia, and hence, nitrogen, is established. The inlet tube allows the permanganate solution to be introduced after distillation. Supplied with an outer Standard Taper 29/42 joint on the flask and an outer Standard Taper 19/38 joint on the Graham condenser. Total height is 410mm; total width is 300mm; maximum O.D. is 97mm. For replacement Graham condensers, see No. 2540-200.
Specifications
IncludesFlask, condenser and inlet tube
Height (Metric)410 mm
For Use With (Application)Determines nitrogen in organic compounds
TypeKjeldahl Nitrogen Distillation Apparatus
MaterialBorosilicate Glass
Height (English)16.14 in.
Capacity16.9 oz., 500 mL
Capacity (Metric)500 mL
Diameter (Metric) Outside97 mm
Length (English) Jacket7.87 in.
Length (Metric) Jacket200 mm
Capacity (English)16.9 oz.
Diameter (English) Outside3.81 in.
Unit SizeCS