Removes interference substances in cyanide quantitative analyses according to ASTM™ D 2036
Have Questions?
Catalog number FIS09-108
Price (MYR)
4,948.00
EA
Add to cart
Price (MYR)
4,948.00
EA
Add to cart
Sample reacts with HS2O4 and MgCl2 to convert any cyanide to hydrocyanic acid (HCN). HCN is distilled and absorbed in NaOH solution, and sample is analyzed for CN.
With standard taper ClearSeal™ grease-free joints and medium frit
Wheaton-33™ low extractable borosilicate glass
Components also available separately; see cat. no. 091081
Specifications
Certifications/ComplianceUSP Type I, ASTM E438 Type I, Class A
For Use With (Application)Ideal for removing interfering substances in quantitative analysis for cyanide
IncludesAbsorber jacket, absorber top, cold finger condenser, cold finger jacket, 1000mL distilling flask, and inlet tube, ClearSeal™ grease-free joints and medium frit