PYREX™ Reusable Petri Dishes: Complete
PYREX™ Reusable Petri Dishes: Complete
Actual product may vary
Fisherbrand
PYREX™ Reusable Petri Dishes: Complete
Flat, clear dishes with beaded edges withstand repeated sterilization (wet or dry)
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Height (Metric)
FIS08-747D100 mm20 mm
FIS08-747B100 mm10 mm
FIS08-747C100 mm15 mm
Catalog number FIS08-747D
Price (MYR)
1,002.00
PK
Add to cart
Diameter (Metric):
100 mm
Height (Metric):
20 mm
Price (MYR)
1,002.00
PK
Add to cart
Flat, clear dishes with beaded edges withstand repeated sterilization (wet or dry)
These flat, clear dishes will withstand repeated sterilization (wet or dry). The edges are beaded to provide greater mechanical strength. The bead also provides a means to equally space the side walls of the bottom and cover, thereby reducing the capillary action of condensed moisture on the sides. They are not affected chemically or thermally by any of the methods commonly employed in laboratories where sterilization is a major factor in routine or in specialized work. The covered dish is not airtight. The tops are marked in blue enamel and the bottoms in white enamel to make sorting easier. Bottoms also have a triangular, enamel reference point for serial dilutions.
Specifications
ColorBlue, White
DisposableReusable
MaterialBorosilicate Glass
ShapeRound
Surface TreatmentNon-treated
TypeCell Culture Petri Dish
EdgesBeaded
No. per Case72/Cs.
No. per Pack12/Pk.
Height (Metric)20 mm
Diameter (Metric)100 mm
Unit SizePK