Check Order Status
Dish Tongs
Actual product may vary
Dish Tongs
Fisherbrand Dish Tongs are made from ASTM™ standard stainless-steel.
Have Questions?
Catalog Number
Length (Metric)
FIS08-725
24 cm
Catalog number
FIS08-725
Price (MYR)
153.00
EA
Add to cart
Length (Metric):
24 cm
Price (MYR)
153.00
EA
Add to cart
Fisherbrand™ Dish Tongs. Our instruments are designed to exceed performance expectations -This is the Fisherbrand Advantage.
Safely handle evaporating dishes from 70 to 250 mL capacity, or similar vessels with 3 to 4.75 in. diameter (7.5 to 12.0 cm)
Raised riveted-joints
Specifications
For Use With (Application)
Safely handling 70 to 250mL evaporating dishes or 3 to 4.75 in. dia. similar vessels
Length (English)
9.5 in.
Length (Metric)
24 cm
Material
Stainless Steel
Type
Dish Tongs