PYREX™ Evaporating Dishes
PYREX™ Evaporating Dishes
Actual product may vary
Fisherbrand
PYREX™ Evaporating Dishes
Withstand temperatures to 230°C (446°F)
Have Questions?
Catalog NumberCapacity (Metric)Diameter (English)Height (English)
FIS08-710A140 mL4.13 in.1.77 in.
Catalog number FIS08-710A
Price (MYR)
930.00
CS
Add to cart
Capacity (Metric):
140 mL
Diameter (English):
4.13 in.
Height (English):
1.77 in.
Price (MYR)
930.00
CS
Add to cart
Withstand temperatures to 230°C (446°F)
These dishes have rugged beaded rims and are of standard medium depth.
Specifications
Bottom ShapeFlat
Capacity (Metric)140 mL
MaterialBorosilicate Glass
ShapeRound
TypeEvaporating Dish
No. per Case6/Cs.
No. per Pack6/Pk.
Temperature Range (English)446°F
Temperature Range (Metric)230°C
Height (Metric)45 mm
Height (English)1.77 in.
Diameter (English)4.13 in.
Diameter (Metric)80 mm
Unit SizeCS