Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO
Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO
Actual product may vary
Fisherbrand
Spectra/Por™ 1 RC Dialysis Membrane Tubing 6000 to 8000 Dalton MWCO
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Length (Metric)
FIS08-670B20.4 mm30.5 m
FIS08-670C25.5 mm30.5 m
FIS08-670D32 mm30.5 m
Catalog number FIS08-670B
Price (MYR)
3,596.00
EA
Add to cart
Diameter (Metric):
20.4 mm
Length (Metric):
30.5 m
Price (MYR)
3,596.00
EA
Add to cart
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Product TypeSpectra/Por Dialysis Tubing
MWCO6 to 8 kDa
MaterialRegenerated Cellulose
Diameter (Metric)20.4 mm
Diameter (English)0.8 in.
Length (English)100 ft.
Length (Metric)30.5 m
Temperature Range (English)39.2°F to 249.8°F
Temperature Range (Metric)4°C to 121°C
QuantityEach
For Use With (Application)Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Unit SizeEA