PYREX™ Class B Mixing Cylinders with Standard Taper Stopper, White Graduations
PYREX™ Class B Mixing Cylinders with Standard Taper Stopper, White Graduations
Actual product may vary
Fisherbrand
PYREX™ Class B Mixing Cylinders with Standard Taper Stopper, White Graduations
Calibrated To Contain
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FIS08-566B25 mL0.2 mL
Catalog number FIS08-566B
Price (MYR)
380.00
EA
Add to cart
Capacity (Metric):
25 mL
Graduations:
0.2 mL
Price (MYR)
380.00
EA
Add to cart
Calibrated 'to contain' in accordance with ASTM E-1272 and ASTM E-542. Useful as a mixing cylinder. Each features the PYREX stopper. Graduations are in durable white enamel. See Cat. No. 3002 for the equivalent cylinder supplied with a Corning test certificate.
For stopper only, see 7650.
Specifications
Base TypeHexagonal
CalibrationTo Contain
Scale ColorWhite
ShapeRound
SterilityNon-sterile
TypeGraduated Mixing Cylinder
Tolerance±0.3 mL
MaterialBorosilicate Glass
Certifications/ComplianceASTM
ClassB
Diameter (English) Outside0.7 in.
Capacity (Metric)25 mL
Diameter (Metric) Outside18 mm
For Use With (Application)Useful as a Mixing Cylinder
Height (English)8.85 in.
Height (Metric)225 mm
IncludesStopper
ScaleSingle
Stopper Number13
Stopper MaterialGlass
Stopper StyleHollow
Tolerance (Metric)±0.3 mL
Closure MaterialGlass
Graduations0.2 mL
AutoclavableAutoclavable
Subdivision0.2 mL
Unit SizeEA