Borosilicate Cylinders with Pouring Lip
Borosilicate Cylinders with Pouring Lip
Actual product may vary
Fisherbrand
Borosilicate Cylinders with Pouring Lip
Heavy-wall construction resists breakage
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FIS08-530J300 mL41 mm325 mm
Catalog number FIS08-530J
Price (MYR)
233.00
EA
Add to cart
Capacity (Metric):
300 mL
Diameter (Metric):
41 mm
Height (Metric):
325 mm
Price (MYR)
233.00
EA
Add to cart
Heavy-wall construction resists breakage
  • Ungraduated
  • Fused round base
  • For use with hydrometers
Specifications
ShapeRound
Base TypeRound
Capacity (English)10.14 oz.
Height (English)12.2 in.
Height (Metric)325 mm
TypeUngraduated Cylinder
Diameter (English)1.61 in.
Diameter (Metric)41 mm
Capacity (Metric)300 mL