Check Order Status
Fused-Quartz Crucibles
Actual product may vary
Fisherbrand
Fused-Quartz Crucibles
Withstand temperatures up to 1250°C (2282°F)
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Diameter (Metric) Outer
Height (Metric)
FIS08-072F
100 mL
6.2cm
6.6cm
FIS08-072D
30 mL
4.3cm
3.8cm
2 Options
Catalog number
FIS08-072F
Price (MYR)
0.00
EA
Add to cart
Capacity (Metric):
100 mL
Diameter (Metric) Outer:
6.2cm
Height (Metric):
6.6cm
Price (MYR)
0.00
EA
Add to cart
Withstand temperatures up to 1250°C (2282°F)
Withstand severe temperature changes
Insoluble in water and acids except hydrofluoric and phosphoric acids
Covers sold separately
Specifications
Color
Clear
Description
High purity fused quartz
Material
Quartz
Max. Temperature (Metric)
1250°C
Thickness (Metric)
2.25mm
Capacity (Metric)
100 mL
Diameter (Metric) Outer
6.2cm
Height (Metric)
6.6cm
Product Type
Crucible