Round Covered Lead Flask Rings
Round Covered Lead Flask Rings
Actual product may vary
Fisherbrand
Round Covered Lead Flask Rings
Stabilize Erlenmeyer flasks immersed in water baths.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) InnerDiameter (English) InnerWeight (English)Weight (Metric)
FIS05-86975.6 cm2.2 in.1.5 lb.680.3 g
FIS05-86965.1 cm2 in.1 lb.453.5 g
FIS05-86954.1 cm1.6 in.0.5 lb.226.7 g
Catalog number FIS05-8697
Price (MYR)
286.00
EA
Add to cart
Diameter (Metric) Inner:
5.6 cm
Diameter (English) Inner:
2.2 in.
Weight (English):
1.5 lb.
Weight (Metric):
680.3 g
Price (MYR)
286.00
EA
Add to cart
Stabilize Erlenmeyer flasks immersed in water baths.
  • Reduces floating, breakage and chemical spills
  • Bright blue coating
  • Chemical-resistant
  • Will not scratch flasks
  • Available in four diameter/weight combinations
Specifications
Product TypeLead Ring
AutoclavableNot Autoclavable
ColorBlue
HoldsFlasks (500 to 2000 mL)
MaterialLead
ShapeRound
ShelvesNone
StackableNo
SterilityNon-sterile
For Use With (Equipment)500 to 2000mL Erlenmeyer flasks immersed in water baths
Length (Metric)102 mm
Width (Metric)46 mm
Height (Metric)102 mm
Diameter (English) Inner2.2 in.
Diameter (Metric) Inner5.6 cm
Weight (English)1.5 lb.
Weight (Metric)680.3 g
Unit SizeEA