Castaloy™ Chain Clamp
Castaloy™ Chain Clamp
Actual product may vary
Fisherbrand
Castaloy™ Chain Clamp
For holding bottles, beakers and other large objects
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS05-745Q05-754 Q Clamp Holder
Catalog number FIS05-745Q
Price (MYR)
-
For Use With (Equipment):
05-754 Q Clamp Holder
For holding bottles, beakers and other large objects
  • Holds 76 to 165mm (3 to 6.5 in.) bottles and large objects
  • Fast, sure way to secure large equipment to rods and lattices
  • Loop steel chain around object; attach to hook on movable slide; tighten slide with large, nonslip knob
  • Holds equipment firmly in place at any angle, variable distance from support
  • Arm is 15cm (6 in.) long and 13mm (0.5 in.) diameter
  • Use with 05-754Q clamp holder
Specifications
Quantity1 Ea.
TypeChain Clamp
MaterialStainless Steel
For Use With (Equipment)05-754 Q Clamp Holder
For Use With (Application)For holding bottles, beakers and other large objects
Length (English) Arm6 in.
Length (Metric) Arm15.24 cm
Unit SizeEA