Kit to culture and evaluate mouse mammary epithelial cells
Have Questions?
Catalog Number
Quantity
FIS05-610
1 Kit
Catalog number FIS05-610
Price (MYR)
607.00
EA
Add to cart
Quantity:
1 Kit
Price (MYR)
607.00
EA
Add to cart
EpiCult™-B (mouse) is a serum-free liquid culture medium optimized for the culture of mouse mammary luminal and myoepithelial cells
It is ideal for the culture and evaluation of mouse mammary epithelial progenitors in the mammary colony-forming unit assay when used in conjunction with an irradiated feeder layer such as NIH 3T3 cells
This medium is also used for enzymatic dissociation of mouse mammary tissue when supplemented with collagenase and hyaluronidase
Addition of Human Recombinant EGF (Catalog #78006), Human Recombinant bFGF (Catalog #78003), and Heparin Solution (Catalog #07980) is required for culturing cells
Specifications
Cell TypeMammary Cells
For Use With (Application)Cell Culture; Colony Assay