IQGAP2 Mouse anti-Human, Mouse, Clone: BB9, MilliporeSigma™
IQGAP2 Mouse anti-Human, Mouse, Clone: BB9, MilliporeSigma™
Actual product may vary
IQGAP2 Mouse anti-Human, Mouse, Clone: BB9, MilliporeSigma™
Mouse Monoclonal Antibody
Have Questions?
Catalog number FIS05-505
Price (MYR)
1,310.00
CS
Add to cart
Price (MYR)
1,310.00
CS
Add to cart
Anti-IQGAP2 Antibody, clone BB9 detects level of IQGAP2 and has been published and validated for use in immunoprecipitation and western blotting.
For Research Use Only
Specifications
AntigenIQGAP2
ApplicationsImmunoprecipitation, Western Blot
ClassificationMonoclonal
CloneBB9
ConjugateUnconjugated
Formulation0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide containing 30% glycerol
Gene Accession No.NM_006633
Gene SymbolsIQGAP2
Host SpeciesMouse
ImmunogenGST-fusion protein corresponding to residues 519-939 of human IQGAP2, purified from E. coli
Purification MethodAscites
Quantity200 μL
Primary or SecondaryPrimary
Target SpeciesHuman, Mouse
FormAscites
IsotypeIgG
Unit SizeCS
Contents & Storage
−20°C for 24 months.