Kimble™ Plain Disposable Borosilicate Glass Tubes
Kimble™ Plain Disposable Borosilicate Glass Tubes
Actual product may vary
Fisherbrand
Kimble™ Plain Disposable Borosilicate Glass Tubes
Suitable for a variety of applications and cell washing procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric)Diameter (Metric)
FIS03341519 mL125 mm16 mm
FIS03-341-04 mL75 mm10 mm
FIS0334104 mL75 mm10 mm
FIS03-341-16 mL75 mm12 mm
FIS0334116 mL75 mm12 mm
FIS03-341-310 mL100 mm13 mm
Catalog number FIS033415
Price (MYR)
1,049.00
CS
Add to cart
Capacity (Metric):
19 mL
Length (Metric):
125 mm
Diameter (Metric):
16 mm
Price (MYR)
1,049.00
CS
Add to cart
Suitable for a variety of applications and cell washing procedures.
  • These premium quality tubes feature sturdy, uniform bottoms and consistent lengths 
  • The 10 x 75 and 12 x 75mm sizes are suitable for cell washing procedures
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • The 6 x 50mm size is often referred to as a “Durham tube”
  • Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination
  • No marking spot
  • Designed from ASTM Specification E890
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
Certifications/ComplianceUSP Type I, ASTM E 438 Type I, Class B
Capacity (Metric)19 mL
Length (Metric)125 mm
MaterialBorosilicate Glass
DisposableSingle-use
For Use WithExcellent chemical resistance against sodium leaching
Label TypeNone
PackagingTray
Diameter (Metric)16 mm
Product TypePlain Tube
Unit SizeCS