Kimble™ KIMAX™ Buret with Funnel Top and PTFE Stopcock
Kimble™ KIMAX™ Buret with Funnel Top and PTFE Stopcock
Actual product may vary
Kimble™ KIMAX™ Buret with Funnel Top and PTFE Stopcock
Kimble™ KIMAX™ Buret with Funnel Top and PTFE Stopcock
Fisherbrand
Kimble™ KIMAX™ Buret with Funnel Top and PTFE Stopcock
Class A
Have Questions?
Catalog NumberCapacity (Metric)
FIS03-76550 mL
Catalog number FIS03-765
Price (MYR)
2,595.00
CS
Add to cart
Capacity (Metric):
50 mL
Price (MYR)
2,595.00
CS
Add to cart
  • Easy to fill with precision bore tubing Durable white scale
  • Permanently marked with individual serial number
  • Inset shows top configuration
Specifications
Capacity (Metric)50 mL
Length (Metric)746 mm
MaterialBorosilicate Glass
Graduations0.1 mL
For Use With (Application)General Purpose Titrations Requiring Traceable Volumetric Accuracy
Length (English)29.37 in.
IncludesNo. 2 Stopcock
ScaleWhite
Stopcock MaterialPTFE
Tolerance±0.05 mL
TypeBore Buret
ClassClass A
Certifications/ComplianceASTM E287, ASTM E438, ASTM E542, ASTM E694, NIST
Unit SizeCS