Schellbach Burets with Straight Standard Taper Glass Stopcock
Schellbach Burets with Straight Standard Taper Glass Stopcock
Actual product may vary
Fisherbrand
Schellbach Burets with Straight Standard Taper Glass Stopcock
Provides accurate readings.
Have Questions?
Catalog NumberCapacity (Metric)
FIS03-740B50 mL
Catalog number FIS03-740B
Price (MYR)
-
Capacity (Metric):
50 mL
Provides accurate readings.
  • Accentuate meniscus for easy reading
  • Accuracy established within the limits of error allowed by NIST™
Specifications
Capacity (Metric)50 mL
MaterialGlass
Graduations0.1 mL
IncludesStraight Stopcock
StopcockStraight
Stopcock MaterialGlass
Tolerance±0.1 mL
TypeSchellbach Buret
Certifications/ComplianceNIST
Unit SizeEA