Check Order Status
Nessler Tube Brushes, Natural black bristle
Actual product may vary
Fisherbrand
Nessler Tube Brushes, Natural black bristle
Fisherbrand™ Nessler Tube Brushes are for use with low- and high-form Nessler tubes.
Have Questions?
Catalog Number
Length (Metric)
Length (Metric) Bristle
For Use With (Equipment)
FIS03-621A
27.94 cm
10 cm
For low-form nessler tubes
Catalog number
FIS03-621A
Price (MYR)
-
Length (Metric):
27.94 cm
Length (Metric) Bristle:
10 cm
For Use With (Equipment):
For low-form nessler tubes
Fisherbrand™ Nessler Tube Brushes are for use with low- and high-form Nessler tubes.
Full bristles to make complete contact with cylinder walls and bottom
03-621A is natural black bristle; others are black nylon
Radial, double-fan tip
Galvanized wire handle
Specifications
Product Type
Nessler Tube Brush
Bristle Material
Natural
Handle Material
Galvanized Wire
Tip Type
Radial, Double Fan Tip
Color
Black
Diameter (English) Bristle
1.75 in.
Diameter (Metric) Bristle
4.4 cm
For Use With (Equipment)
For low-form nessler tubes
Length (English)
11 in.
Length (English) Bristle
4 in.
Length (Metric)
27.94 cm
Length (Metric) Bristle
10 cm
Size
Medium