PYREX™ Gas Washing, Tall-Form, Fritted Cylinder Bottles
PYREX™ Gas Washing, Tall-Form, Fritted Cylinder Bottles
Actual product may vary
Fisherbrand
PYREX™ Gas Washing, Tall-Form, Fritted Cylinder Bottles
Tall design permits thorough washing.
Have Questions?
Catalog number FIS03-040D
Price (MYR)
2,891.00
CS
Add to cart
Price (MYR)
2,891.00
CS
Add to cart
Stopper is equipped with a 12 mm diameter fritted cylinder instead of a disc as the distributor.
Specifications
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
MaterialBorosilicate Glass
TypeGas Washing Bottle
Includes0.47 in. extra coarse fritted cylinder
Pore Size170 to 220 μm
PorosityExtra Coarse
Standard Taper Joint29/42
SterilityNon-sterile
Diameter (English) Outside2.007 in.
Diameter (Metric) Outside5.1 cm
For Use With (Application)Provide an inexpensive but effective method for washing and drying gases
Quantity1/Cs.
Unit SizeCS