Kimble™ Amber Dropping Bottles
Kimble™ Amber Dropping Bottles
Actual product may vary
DWK Life Sciences
Kimble™ Amber Dropping Bottles
With 1mL dropper
Have Questions?
Catalog NumberCapacity (Metric)Includes
FIS02-991C60 mLGlass Dropper
Catalog number FIS02-991C
Price (MYR)
708.00
CS
Add to cart
Capacity (Metric):
60 mL
Includes:
Glass Dropper
Price (MYR)
708.00
CS
Add to cart
With 1mL dropper
  • ASTM No. 438, Type II
Specifications
Capacity2.02 oz., 60 mL
Capacity (Metric)60 mL
Capacity (English)2.02 oz.
ColorAmber
Diameter (English)1.53 in.
IncludesGlass Dropper
MaterialBorosilicate Glass
Dropper Capacity1 mL
Height (Metric)118 mm
Product TypeAmber Dropping Bottle
Width (English)1.53 in.
Height (English)4.64 in.
Diameter (Metric)39 mm
Through the Closure DispensingYes
Unit SizeCS