Amber Glass Dropping Bottles
Amber Glass Dropping Bottles
Actual product may vary
Amber Glass Dropping Bottles
Ideal for light-sensitive chemicals
Have Questions?
Catalog NumberCapacity (Metric)
FIS02-983B60 mL
Catalog number FIS02-983B
Price (MYR)
1,167.00
CS
Add to cart
Capacity (Metric):
60 mL
Price (MYR)
1,167.00
CS
Add to cart
Ideal for light-sensitive chemicals
  • Amber glass protects light-sensitive chemicals
Specifications
Capacity2.02 oz., 60mL
Capacity (Metric)60 mL
Capacity (English)2.02 oz.
ColorAmber
MaterialGlass
SterilityNon-sterile
Dropper Capacity60 mL
ShapeRound
Product TypeDropping Bottle
Closure MaterialPolypropylene
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeCS