Hemato-Seal™ Capillary Tube Sealant
Hemato-Seal™ Capillary Tube Sealant
Actual product may vary
Fisherbrand
Hemato-Seal™ Capillary Tube Sealant
Nondrying, nonhardening, nontoxic compound for sealing hematocrit and other blood collecting tubes
Have Questions?
Catalog NumberQuantity
FIS02-67810 Pk.
Catalog number FIS02-678
Price (MYR)
298.00
PK
Add to cart
Quantity:
10 Pk.
Price (MYR)
298.00
PK
Add to cart
Nondrying, nonhardening, nontoxic compound for sealing hematocrit and other blood collecting tubes
  • Light color provides sharp contrast for easy reading
  • Excellent retention during centrifugation
Specifications
Product TypeCapillary Tube Sealant
For Use With (Application)Sealing hematocrit and other Blood collecting tubes, provides excellent retention during centrifugation
For Use With (Equipment)Hemato-Seal™ capillary tube
Quantity10 Pk.
Unit SizePK