PYREX™ Tall-Form Berzelius Beakers with Spout, Graduated
PYREX™ Tall-Form Berzelius Beakers with Spout, Graduated
Actual product may vary
Fisherbrand
PYREX™ Tall-Form Berzelius Beakers with Spout, Graduated
Graduated to indicate approximate content
Have Questions?
Catalog NumberCapacity (Metric)
FIS02-546E500 mL
Catalog number FIS02-546E
Price (MYR)
466.00
PK
Add to cart
Capacity (Metric):
500 mL
Price (MYR)
466.00
PK
Add to cart
Corning™ PYREX™ Tall-Form Berzelius Beakers are graduated to indicate approximate content.
A tall beaker with spout to facilitate pouring. For convenience, these beakers are graduated to indicate their approximate content.
Specifications
Capacity (English)16.9 oz.
Diameter (English) Outer2.95 in.
Diameter (Metric) Outer7.5 cm
Capacity (Metric)500 mL
MaterialBorosilicate Glass
For Use With (Application)Convenient for Easy Pouring
Graduation Range50 to 450 mL
Height (English)5.34 in.
Height (Metric)13.6 cm
IncludesSpout
Increments50 mL
Label TypeMarking Spot
Quantity6 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960
ScaleWhite Enamel Scale
StyleBerzelius
Unit SizePK