Kimble™ KIMAX™ Griffin Beakers
Kimble™ KIMAX™ Griffin Beakers
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Griffin Beakers
With thick, beaded, slightly flared top
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-539P1000 mL
FIS02-539C10 mL
FIS02-539G50 mL
FIS02-539K250 mL
Catalog number FIS02-539P
Price (MYR)
732.00
PK
Add to cart
Capacity (Metric):
1000 mL
Price (MYR)
732.00
PK
Add to cart
With thick, beaded, slightly flared top
  • With pouring spout
  • Matte-finish enameled marking spot
  • 20 to 4000mL sizes have white scales for approximate volume
  • Accurate to ±5%
  • Uniform, sturdy walls
Specifications
Accuracy±5%
Capacity (English)33.81 oz.
Capacity (Metric)1000 mL
MaterialBorosilicate Glass
For Use With (Application)For Measuring and Mixing Liquids
Graduation Range50 to 1000 mL
IncludesSpout
Increments50 mL
Label TypeMatte Finish Enamel Marking
Quantity6 Pk.
TypeBeaker
GraduatedYes
Certifications/ComplianceASTM E960, Type I
ScaleDual White Scale
StyleGriffin Beaded Top
Unit SizePK