Autoclavable Waste Bags: Plain
Autoclavable Waste Bags: Plain
Actual product may vary
Autoclavable Waste Bags: Plain
Plain style (without print).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (L x W)Capacity (English)
FIS01-830CRed23 x 19 in. (58 x 48 cm)7 to 10 gal.
FIS01-826-2Clear10 x 10 in.1 to 2 gal.
FIS01-826-4Clear10 x 15 in. (25 x 38 cm)1 to 2 gal.
FIS01-826-5Clear24 x 12 in. (61 x 30 cm)2 to 3 gal.
FIS01-830ARed12 x 8 in. (30 x 20 cm)1 to 2 gal.
FIS01-830BRed19 x 14 in. (48 x 36 cm)3 to 4 gal.
Catalog number FIS01-830C
Price (MYR)
484.00
CS
Add to cart
Color:
Red
Dimensions (L x W):
23 x 19 in. (58 x 48 cm)
Capacity (English):
7 to 10 gal.
Price (MYR)
484.00
CS
Add to cart
Plain style bags
  • Maximum temperature, 140°C (285°F); for use at 134°C (274°F)
  • 2mil thick
  • Supplied with autoclavable elastic steam-venting closures
Specifications
Certifications/ComplianceOSHA, 29 CFR1910.1030
Closure TypeElastic
Capacity (English)7 to 10 gal.
ColorRed
Dimensions (L x W)23 x 19 in. (58 x 48 cm)
Length (Metric)58 cm
Width (Metric)48 cm
AutoclavableAutoclavable
Width (English)19 in.
Length (English)23 in.
Thickness2 mil
MaterialPolypropylene
Temperature (Metric)140°C
Unit SizeCS