Wheaton™ Prescored Gold-Band™ Clear Ampules
Wheaton™ Prescored Gold-Band™ Clear Ampules
Actual product may vary
DWK Life Sciences
Wheaton™ Prescored Gold-Band™ Clear Ampules
Prescored to eliminate need for filing
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (English) OuterHeight (English)
FIS01-215D10 mL0.74 in.4.21 in.
FIS01-215A1 mL0.41 in.2.63 in.
FIS01-215C5 mL0.64 in.3.3 in.
FIS01-215B2 mL0.47 in.2.95 in.
Catalog number FIS01-215D
Price (MYR)
781.00
CS
Add to cart
Capacity (Metric):
10 mL
Diameter (English) Outer:
0.74 in.
Height (English):
4.21 in.
Price (MYR)
781.00
CS
Add to cart
Prescored to eliminate need for filing
  • Low-extractable borosilicate glass
  • Gold-Band feature indicates prescoring
  • Stems can be pull sealed or tip sealed
Specifications
Capacity (Metric)10 mL
MaterialBorosilicate Glass
Product TypePrescored Ampule
ColorClear
Diameter (Metric) Outer1.9 cm
Diameter (English) Outer0.74 in.
Height (Metric)10.7 cm
Height (English)4.21 in.
Certifications/ComplianceUSP Type 1, ASTM Type I, Class A
Unit SizeCS