SANOclav™ La-MCS Autoclave
Product Image
Fisherbrand
SANOclav™ La-MCS Autoclave
For sterilization in tightened steam and water. Adolf Wold SANOclav™ SANOclav La-MCS Autoclave is fitted with safety valve, over temperature protection, steam outlet valve, pressure gauge and microprocessor controller.
Have Questions?
Catalog NumberTemperature (Metric)Operating Pressure
FIS01-002151°C4 bar max.
Catalog number FIS01-002
Price (MYR)
752.00
EA
Add to cart
Temperature (Metric):
151°C
Operating Pressure:
4 bar max.
Price (MYR)
752.00
EA
Add to cart
  • Volume of the pressure chamber: 20 litres
  • Lid and chamber made of stainless steel
  • Fitted with safety valve, over temperature protection, steam outlet valve, pressure gauge and microprocessor controller
  • Electrical heating 230 volts / 1500 watts
Options: Must be built in in factory
  • Connection to a PC: USB connection with program to record the time and the temperature on an external personal computer
  • Liquid sensor: Flexible PT 100 sensor for liquids (when the liquid sensor is on, a pre- and post- warming temperature of 60°C will be held
  • Safety lock: Autoclave locks with temperatures over 97°C or over 80°C with liquids (only for type La-MCS-203 / 204 / VA)
  • Filter system: To filter the vapor
Specifications
Controller TypeMicroprocessor
DescriptionBenchtop autoclave sterilizer with safety lock
Capacity (Metric)20 L
Depth (Metric) Chamber360 mm
Height (Metric) Exterior530 mm
Length (Metric)390 mm
ModelLa-MCS-204
Product TypeBenchtop Autoclave
Temperature (Metric)151°C
Weight (Metric)27 kg
Electrical Requirements230V / 1500W
FeaturesSafety valve, over-temperature protection, steam outlet, pressure gauge, microprocessor controller
Height (Metric) Interior400 mm
Operating Pressure4 bar max.
Quantity1 Ea.
Sterilization MethodSteam
Wattage1500 w
Unit SizeEA