Hose Adapter Kit for Rotary Vane Vacuum Pumps
Product Image
Hose Adapter Kit for Rotary Vane Vacuum Pumps
Use this hose adaptor kit with all Fisherbrand rotary van vacuum pumps. Fisherbrand™ Hose Adapter Kits for Rotary Vane Vacuum Pumps include stainless steel tubing adaptors and clamps for vacuum tight connections.
Have Questions?
Catalog number FIL330060
Price (MYR)
3,699.00
EA
Add to cart
Price (MYR)
3,699.00
EA
Add to cart
  • Vacuum hose adapter kit
  • For Fisherbrand rotary vane vacuum pumps
  • Stainless steel tubing adaptors and clamps for vacuum tight connections
  • 1.5m of thick walled rubber hose in Sizes DN10 and DN20
Specifications
IncludesDN10 & DN20 hose, Stainless Steel adapters and clamps
For Use WithAll Fisherbrand vacuum pumps
Pump TypeHose Adapter Kit for Rotary Vane Vacuum Pumps
Unit SizeEA