Kit 5 Freeze dryer adapter kit, for Chemvac & Vacuum concentrator Adapter kit, DN 16 KF
Product Image
Kit 5 Freeze dryer adapter kit, for Chemvac & Vacuum concentrator Adapter kit, DN 16 KF
For rotary vane pump DN 16 KF
Have Questions?
Catalog number FIL330044
Price (MYR)
1,596.00
EA
Add to cart
Price (MYR)
1,596.00
EA
Add to cart
Vacuum concentrator kit includes:
  • KF16 flange adapter
  • 1.5m PVC vacuum hose
  • Clamps, centring rings and adapters
Specifications
DescriptionDN 16 KF
For Use With (Equipment)Rotary vane pump DN 16 KF
Unit SizeEA