Oil Mist Separator for CRVpro Direct Drive Rotary Vane Vacuum Pump
Oil Mist Separator for CRVpro Direct Drive Rotary Vane Vacuum Pump
Actual product may vary
Oil Mist Separator for CRVpro Direct Drive Rotary Vane Vacuum Pump
For use with CRVpro Direct Drive Rotary Vane Vacuum Pump
Have Questions?
Catalog number FIL320015
Price (MYR)
2,861.00
EA
Add to cart
Price (MYR)
2,861.00
EA
Add to cart
  • Outlet oil separator removes oil particles from exhaust stream, minimizing oil loss to atmosphere
Specifications
DescriptionOutlet Oil Mist Separator
For Use With (Application)Removes oil particles from exhaust stream
For Use WithRotary vane pump
Unit SizeEA