Check Order Status
Zinc Acetate Dihydrate, Certified AR for Analysis, Fisher Chemical™
Element Search
Structure Search
Zinc Acetate Dihydrate, Certified AR for Analysis, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCZ070053
also known as FGCZ/0700/53
500 g
Catalog number
FGCZ070053
also known as FGCZ/0700/53
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Melting Point
237°C
CAS
5970-45-6
Color
White
pH
6.0 to 7.0
Physical Form
Solid
View more