Check Order Status
Zinc, Certified AR for Analysis, Granulated Metal, Fisher Chemical™
Element Search
Structure Search
Zinc, Certified AR for Analysis, Granulated Metal, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCZ030053
also known as FGCZ/0300/53
500 g
Catalog number
FGCZ030053
also known as FGCZ/0300/53
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Color
Gray
CAS
7440-66-6
Melting Point
419°C
Boiling Point
907°C
Molecular Formula
Zn
View more
Zinc, Certified AR for Analysis, Granulated Metal