Check Order Status
O-Xylene, Pure, Fisher Chemical™
Element Search
Structure Search
O-Xylene, Pure, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCX030017
also known as FGCX/0300/17
2.5 L
Catalog number
FGCX030017
also known as FGCX/0300/17
Price (MYR)
-
Quantity:
2.5 L
Request bulk or custom format
Specifications
Melting Point
-25°C
CAS
95-47-6
Color
Colorless
Boiling Point
143°C
Quantity
2.5 L
View more