Water, Optima™ LC/MS Grade, Fisher Chemical™
Water, Optima™ LC/MS Grade,  Fisher Chemical™
Water, Optima™ LC/MS Grade,  Fisher Chemical™
Water, Optima™ LC/MS Grade,  Fisher Chemical™
Fisher Chemical
Water, Optima™ LC/MS Grade, Fisher Chemical™
Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max. | CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Catalog number FGCW6-1
Price (MYR)
406.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
406.00
EA
Add to cart
Specifications
Absorbance≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm
Chemical Name or MaterialWater
pH7
CAS7732-18-5
ColorColorless
View more
Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 1L, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, <-0.01 AU at 210nm, <-0.01 AU at 230nm, <-0.01 AU at 240nm, <-0.01 AU at 220nm, 10 ppb max., 100 deg.C, 20 ppb max.