Water, for UHPLC Gradient Grade Analysis, Fisher Chemical™
Water, for UHPLC Gradient Grade Analysis, Fisher Chemical™
Water, for UHPLC Gradient Grade Analysis, Fisher Chemical™
Water, for UHPLC Gradient Grade Analysis, Fisher Chemical™
Water, for UHPLC Gradient Grade Analysis, Fisher Chemical™
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Catalog number FGCW0120PB17
also known as FGCW/0120/PB17
Price (MYR)
339.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
339.00
EA
Add to cart
Specifications
CAS7732-18-5
ColorColorless
Boiling Point100°C
Molecular FormulaH2O
MDL Number11332
View more
  • Efficiency: lot-to-lot consistency ensures reproducible results
  • Low acidity/alkalinity levels avoid interactions with samples
  • Peak shape: low baseline noise at 210nm and 254nm improves peak shape
  • Speed: filtered to 0.1μm, allowing analysis to be performed faster at higher pressures with less risk of pump blockages
  • Sensitivity: interference-free with outstandingly high UV transmission for improved resolution and sensitivity