Check Order Status
Tween™ 20, Pure, Fisher Chemical
Element Search
Structure Search
Tween™ 20, Pure, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCT420660
also known as FGCT/4206/60
1 kg
Catalog number
FGCT420660
also known as FGCT/4206/60
Price (MYR)
436.00
BOT
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
436.00
BOT
Add to cart
Specifications
Color
Amber
CAS
9005-64-5
Physical Form
Very viscous Liquid
Boiling Point
100°C
pH
6
View more