Check Order Status
Trypsin, Pure, From Beef Pancreas, Fisher Chemical
Actual product may vary
Trypsin, Pure, From Beef Pancreas, Fisher Chemical
Have Questions?
Catalog Number
Quantity
FGCT376048
also known as FGCT/3760/48
100 g
Catalog number
FGCT376048
also known as FGCT/3760/48
Price (MYR)
-
Quantity:
100 g
Specifications
CAS
9002-07-7
Color
White
Format
Crystalline Powder Solid
Molecular Weight (g/mol)
725.80
Quantity
100 g