Triton™ X-100, Extra Pure, SLR, Fisher Chemical™
Triton™ X-100, Extra Pure, SLR, Fisher Chemical™
Actual product may vary
Triton™ X-100, Extra Pure, SLR, Fisher Chemical™
Triton™ X-100, Extra Pure, SLR, Fisher Chemical™
Triton™ X-100, Extra Pure, SLR, Fisher Chemical™
For solubilization and permeabilization of membranes
Have Questions?
Catalog NumberQuantity
FGCT375117
also known as FGCT/3751/17
2.5 L
Catalog number FGCT375117
also known as FGCT/3751/17
Price (MYR)
-
Quantity:
2.5 L
For solubilization and permeabilization of membranes
This nonionic detergent is used to solubilize membranes, permeabilize unfixed eukaryotic membranes, and is a component of lysis buffers for DNA extraction.
Specifications
ColorClear Yellow
CAS9002-93-1
Molecular FormulaC34H62O11
Formula Weight646.86g/mol
Quantity2.5 L