Check Order Status
N-Tris(Hydroxymethyl)Methyl-2-Aminoethanesulfonic Acid, for HPLC, Biological Buffer, Fisher Chemical
Element Search
Structure Search
N-Tris(Hydroxymethyl)Methyl-2-Aminoethanesulfonic Acid, for HPLC, Biological Buffer, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FGCT372048
also known as FGCT/3720/48
100 g
Plastic powder jar
Catalog number
FGCT372048
also known as FGCT/3720/48
Price (MYR)
-
Quantity:
100 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS
7365-44-8
Melting Point
223°C
Color
White
pH
4.0 to 4.8
Packaging
Plastic powder jar
View more