Isooctane, Optima™ for HPLC and GC, Fisher Chemical™
Isooctane, Optima™ for HPLC and GC, Fisher Chemical™
Isooctane, Optima™ for HPLC and GC, Fisher Chemical™
Isooctane, Optima™ for HPLC and GC, Fisher Chemical™
Isooctane, Optima™ for HPLC and GC, Fisher Chemical™
High purity and versatile solvent suitable for HPLC and pesticide residue analysis
Have Questions?
Catalog NumberQuantityPackaging
FGCT360617
also known as FGCT/3606/17
2.5 LAmber Glass Bottle
Catalog number FGCT360617
also known as FGCT/3606/17
Price (MYR)
690.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Price (MYR)
690.00
EA
Add to cart
Specifications
Melting Point-107°C
CAS540-84-1
ColorColorless
Boiling Point98°C
Quantity2.5 L
View more
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only