Check Order Status
Triethylamine, for HPLC, Fisher Chemical™
Actual product may vary
Triethylamine, for HPLC, Fisher Chemical™
C6H15N, CAS Number-121-44-8, 250mL, 90 deg.C, CHEBI:35026, Colorless, 101.19g/mol, ZMANZCXQSJIPKH-UHFFFAOYSA-N, N,N-diethylethanamine, -115 deg.C, 9051, 101.193, Amber glass bottle, 12.4, Liquid, 8471, CCN(CC)CC, 3.5, 69mbar at 20 deg.C, 0.36 mPaS at 20 deg.C | CAS: 121-44-8 | C6H15N | 101.193 g/mol
Have Questions?
Catalog Number
Quantity
FGCT320307
also known as FGCT/3203/07
250 mL
Catalog number
FGCT320307
also known as FGCT/3203/07
Price (MYR)
313.00
BOT
Add to cart
Quantity:
250 mL
Price (MYR)
313.00
BOT
Add to cart
C6H15N, CAS Number-121-44-8, 250mL, 90 deg.C, CHEBI:35026, Colorless, 101.19g/mol, ZMANZCXQSJIPKH-UHFFFAOYSA-N, N,N-diethylethanamine, -115 deg.C, 9051, 101.193, Amber glass bottle, 12.4, Liquid, 8471, CCN(CC)CC, 3.5, 69mbar at 20 deg.C, 0.36 mPaS at 20 deg.C
Specifications
Packaging
Amber glass bottle
CAS
121-44-8
Vapor Pressure
69mbar at 20°C
Viscosity
0.36 mPaS at 20°C
Vapor Density
3.5
Physical Form
Liquid
Molecular Formula
C6H15N
MDL Number
9051
Color
Colorless
pH
12.4
Melting Point
-115°C
Boiling Point
90°C
InChI Key
ZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILES
CCN(CC)CC
IUPAC Name
N,N-diethylethanamine
Molecular Weight (g/mol)
101.193
PubChem CID
8471
Quantity
250 mL
ChEBI
CHEBI:35026
Formula Weight
101.19g/mol