Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™
Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™
Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™
Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™
Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™
C2HCl3O2, CAS Number-76-03-9, trichloressigsaeure, konesta, amchem grass killer, acido tricloroacetico, acide trichloracetique, aceto-caustin, acetic acid, trichloro, trichloroethanoic acid, trichloroacetic acid, trichloroacetic acid, 500g, 196 deg.C, CHEBI:30956, White, 163.39g/mol | CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
500 g
Catalog number FGCT300060
also known as FGCT/3000/60
Price (MYR)
775.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
775.00
EA
Add to cart
Specifications
pH1.2
CAS76-03-9
Melting Point52°C
Boiling Point196°C
ColorWhite
View more
C2HCl3O2, CAS Number-76-03-9, trichloressigsaeure, konesta, amchem grass killer, acido tricloroacetico, acide trichloracetique, aceto-caustin, acetic acid, trichloro, trichloroethanoic acid, trichloroacetic acid, trichloroacetic acid, 500g, 196 deg.C, CHEBI:30956, White, 163.39g/mol