Check Order Status
Toluene, 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Element Search
Structure Search
Toluene, 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
FGCT232317
also known as FGCT/2323/17
2.5 L
Catalog number
FGCT232317
also known as FGCT/2323/17
Price (MYR)
-
Quantity:
2.5 L
Request bulk or custom format
Specifications
Melting Point
-95°C
CAS
108-88-3
Color
Colorless
Boiling Point
111°C
Quantity
2.5 L
View more