Titanium(III) Sulfate, Technical, ≈15% (w/v) solution in diluted sulfuric acid
Titanium(III) Sulfate, Technical, ≈15% (w/v) solution in diluted sulfuric acid
Actual product may vary
Titanium(III) Sulfate, Technical, ≈15% (w/v) solution in diluted sulfuric acid
Titanium(III) Sulfate, Technical, ≈15% (w/v) solution in diluted sulfuric acid
Titanium(III) Sulfate, Technical, ≈15% (w/v) solution in diluted sulfuric acid
O12S3Ti2, CAS Number-10343-61-0, 69hci1s2xi, acmc-1bolh, dititanium tris sulphate, sulfuric acid, titanium 3+ salt 3:2, titan iii-sulfat, titanium 3+ sulfate 2:3, titanium iii sulfate, titanium sesquisulfate, unii-69hci1s2xi, 500mL, Purple, SOBXOQKKUVQETK-UHFFFAOYSA-H, titanium(3+);trisulfate, 36317
Have Questions?
Catalog NumberQuantity
FGCT2000PB08
also known as FGCT/2000/PB08
500 mL
Catalog number FGCT2000PB08
also known as FGCT/2000/PB08
Price (MYR)
-
Quantity:
500 mL
O12S3Ti2, CAS Number-10343-61-0, 69hci1s2xi, acmc-1bolh, dititanium tris sulphate, sulfuric acid, titanium 3+ salt 3:2, titan iii-sulfat, titanium 3+ sulfate 2:3, titanium iii sulfate, titanium sesquisulfate, unii-69hci1s2xi, 500mL, Purple, SOBXOQKKUVQETK-UHFFFAOYSA-H, titanium(3+);trisulfate, 36317
Specifications
ColorPurple
CAS10343-61-0
Molecular FormulaO12S3Ti2
MDL Number36317
Synonymtitanium sesquisulfate, titanium iii sulfate, unii-69hci1s2xi, 69hci1s2xi, dititanium tris sulphate, titanium 3+ sulfate 2:3, titan iii-sulfat, acmc-1bolh, sulfuric acid, titanium 3+ salt 3:2
InChI KeySOBXOQKKUVQETK-UHFFFAOYSA-H
SMILES[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Ti+3].[Ti+3]
IUPAC Nametitanium(3+);trisulfate
Molecular Weight (g/mol)383.902
PubChem CID165866
PackagingPlastic Coated Glass bottle
pH1
Quantity500 mL
Physical FormLiquid