Check Order Status
Tetrachloroethylene, UV Spectroscopy, Fisher Chemical™
Element Search
Structure Search
Tetrachloroethylene, UV Spectroscopy, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FGCT0605PB17
also known as FGCT/0605/PB17
2.5 L
Plastic Coated Glass bottle
Catalog number
FGCT0605PB17
also known as FGCT/0605/PB17
Price (MYR)
1,027.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Request bulk or custom format
Price (MYR)
1,027.00
EA
Add to cart
Specifications
CAS
127-18-4
Melting Point
-22°C
Color
Colorless
Boiling Point
120°C
Packaging
Plastic Coated Glass bottle
View more