Check Order Status
Sulfuric Acid Min 90% d=1.83, SLR, Extra Pure, for Milk Testing, (Gerber Test), Fisher Chemical™
Element Search
Structure Search
Sulfuric Acid Min 90% d=1.83, SLR, Extra Pure, for Milk Testing, (Gerber Test), Fisher Chemical™
H2O4S, CAS Number-7664-93-9, electrolyte acid, dipping acid, dihydrogen sulfate, acide sulfurique, acidum sulfuricum, battery acid, sulphuric acid, mattling acid, oil of vitriol, vitriol brown oil, 2.5 L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid
Have Questions?
Catalog Number
Quantity
Packaging
FGCS9360PB17
also known as FGCS/9360/PB17
2.5 L
HDPE Plastic Bottle
Catalog number
FGCS9360PB17
also known as FGCS/9360/PB17
Price (MYR)
419.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
HDPE Plastic Bottle
Request bulk or custom format
Price (MYR)
419.00
EA
Add to cart
Specifications
pH
1
Color
Colorless
CAS
7664-93-9
Melting Point
10°C
Boiling Point
290°C
View more
H2O4S, CAS Number-7664-93-9, electrolyte acid, dipping acid, dihydrogen sulfate, acide sulfurique, acidum sulfuricum, battery acid, sulphuric acid, mattling acid, oil of vitriol, vitriol brown oil, 2.5 L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid