Sulfuric Acid Min 95% d=1.83, SLR, Extra Pure, Low In Nitrogen, Fisher Chemical™
Sulfuric Acid Min 95% d=1.83, SLR, Extra Pure, Low In Nitrogen, Fisher Chemical™
Sulfuric Acid Min 95% d=1.83, SLR, Extra Pure, Low In Nitrogen, Fisher Chemical™
Sulfuric Acid Min 95% d=1.83, SLR, Extra Pure, Low In Nitrogen, Fisher Chemical™
Sulfuric Acid Min 95% d=1.83, SLR, Extra Pure, Low In Nitrogen, Fisher Chemical™
H2O4S, CAS Number-7664-93-9, acidum sulfuricum, acide sulfurique, battery acid, dihydrogen sulfate, mattling acid, electrolyte acid, vitriol brown oil, oil of vitriol, sulphuric acid, dipping acid, 2.5L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid
Have Questions?
Catalog NumberQuantity
FGCS9200PB17
also known as FGCS/9200/PB17
2.5 L
Catalog number FGCS9200PB17
also known as FGCS/9200/PB17
Price (MYR)
436.00
EA
Add to cart
Quantity:
2.5 L
Request bulk or custom format
Price (MYR)
436.00
EA
Add to cart
Specifications
pH1
ColorColorless
CAS7664-93-9
Melting Point10°C
Boiling Point290°C
View more
H2O4S, CAS Number-7664-93-9, acidum sulfuricum, acide sulfurique, battery acid, dihydrogen sulfate, mattling acid, electrolyte acid, vitriol brown oil, oil of vitriol, sulphuric acid, dipping acid, 2.5L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid