Check Order Status
Chemical Binder Spillage Treatment, Pure, Fisher Chemical™
Actual product may vary
Chemical Binder Spillage Treatment, Pure, Fisher Chemical™
Universally applicable chemical binder suitable for use with all chemical spills.
Have Questions?
Catalog Number
Packaging Quantity
FGCS743553
also known as FGCS/7435/53
500 g
Catalog number
FGCS743553
also known as FGCS/7435/53
Price (MYR)
554.00
EA
Add to cart
Packaging Quantity:
500 g
Price (MYR)
554.00
EA
Add to cart
Universally applicable chemical binder suitable for use with all chemical spills.
Binds with liquids with a volume up to 75 times its own weight
Does not eliminate the hazards associated with a spillage and the residue must be handled and disposed of according to the substance bound
Specifications
Color
Green
Packaging Quantity
500 g